Learn More
ACROBiosystems Biotinylated SARS-CoV-2 (COVID-19) S1 protein, His,Avitag™ (MALS verified)
Supplier:  ACROBiosystems S1NC82E91MG
Encompass_Preferred
Recombinant Protein;1MG;Biotinylated SARS-CoV-2 S1 protein, His,Avitag (S1N-C82E9) is expressed from human 293 cells (HEK293).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 78.6 kDa. The protein migrates as 100-150 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human ACE2, Fc Tag (Cat. No. AC2-H5257) at 2 μg/mL (100 μL/well) can bind Biotinylated SARS-CoV-2 S1 protein, His,Avitag (Cat. No. S1N-C82E9) with a linear range of 20-312 ng/mL (QC tested).
Catalog No. 50-205-4847
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.