Learn More
ACROBiosystems ACROBiosystems Biotinylated SARS-CoV-2 S protein (L18F, T20N, P26S, D138Y, R190S, K417T, E484K, N501Y, D614G, H655Y, T1027I, V1176F) trimer, His,Avitag™ (MALS verified)
Supplier: ACROBiosystems SPNC82E625UG
Recombinant Protein;25UG;Biotinylated SARS-CoV-2 S protein trimer, His,Avitag (SPN-C82E6) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Pro 1213 (Accession # QHD43416.1). The recombinant protein is expressed from human 293 cells (HEK293) with T4 fibritin trimerization motif and a polyhistidine tag at the C-terminus. Proline substitutions (F817P/ A892P/ A899P/ A942P/ K986P/ V987P) and alanine substitutions (R683A and R685A) are introduced to stabilize the trimeric prefusion state of SARS-CoV-2 S protein and abolish the furin cleavage site, respectively. L18F/ T20N/ P26S/ D138Y/ R190S/ K417T/ E484K/ N501Y/ D614G/ H655Y/ T1027I/ V1176F) mutations were identified on the spike protein in the SARS-CoV-2 variant (known as P.1) which emerged in Brazil.;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 139.8 kDa. The protein migrates as 160-200 kDa under reducing (R) condition (SDS-PAGE) due to glycosylat
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.