Learn More
ACROBiosystems ACROBiosystems Biotinylated SARS-CoV-2 Spike Trimer (P26S, HV69-70del, V126A, Y144del, LLA241-243del, H245Y, S477N, E484K, D614G, P681H, T1027I, D1118H), His,Avitag™ (MALS verified)
Supplier:  ACROBiosystems SPNC82ED25UG
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated SARS-CoV-2 Spike Trimer, His,Avitag (SPN-C82Ed) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Pro 1213 (Accession # QHD43416.1). The mutations (P26S, HV69-70del, V126A, Y144del, LLA241-243del, H245Y, S477N, E484K, D614G, P681H, T1027I, D1118H) were identified in the SARS-CoV-2 variants which emerged in Europe (known as B.1.620). Proline substitutions (F817P, A892P, A899P, A942P, K986P, V987P) and alanine substitutions (R683A and R685A) are introduced to stabilize the trimeric prefusion state of SARS-CoV-2 S protein and abolish the furin cleavage site, respectively. ;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 139.1 kDa. The protein migrates as 160-200 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated SARS-CoV-2 Spike Trimer, His,Avitag (Cat. No. SPN-C82Ed) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N
Catalog No. 50-210-6982
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.