Learn More
ACROBiosystems ACROBiosystems Biotinylated SARS-CoV-2 Spike RBD (L452R), His,Avitag™ (MALS verified)
Supplier:  ACROBiosystems SPDC82E325UG
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated SARS-CoV-2 Spike RBD (L452R), His,Avitag (SPD-C82E3) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (L452R)). The L452R mutation was identified in the SARS-CoV-2 variant (known as B.1.427/B.1.429) which emerged in California, the U.S..;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 28.3 kDa. The protein migrates as 30-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human ACE2, Fc Tag (Cat. No. AC2-H5257) at 1 μg/mL (100 μL/well) can bind Biotinylated SARS-CoV-2 Spike RBD, His,Avitag (Cat. No. SPD-C82E3) with a linear range of 2-63 ng/mL (QC tested).;Immobilized Biotinylated SARS-CoV-2 Spike RBD, His,Avitag (Cat. No. SPD-C82E3) at 1 μg/mL (100 μL/well) on streptavidin precoated (0.5 μg/well) plate can bind Human ACE2, Fc Tag (Cat. No. AC2-H5257) with a linear range of 0.2-2 ng/mL (Routinely te
Catalog No. 50-210-7186
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.