Learn More
ACROBiosystems ACROBiosystems Biotinylated Monoclonal Anti-DNP antibody, Mouse IgG1 Avitag™ Isotype Control
Supplier:  ACROBiosystems DNPBM19025TESTS
Encompass_Preferred
Antibody;25TESTS ;Mouse IgG1/kappa;Specifically reacts with DNP (Dinitrophenyl) and DNP conjugated proteins.;Dinitrophenyl (DNP) is a hapten that is not normally expressed in the target tissue. Therefore, anti-DNP antibody has no relevant specificity to a target antigen, and can be used as isotype control antibody to differentiate non-specific background signal from specific antibody signal in various in vitro and in vivo studies. Biotinylated Monoclonal Anti-DNP antibody, Mouse IgG1 Isotype Control was purified from HEK293 cell culture. ;This protein . ;2e5 of anti-CD19 CAR-293 cells were stained with 100 μL of 1:50 dilution (2 μL stock solution in 100 μL FACS buffer) Biotinylated Monoclonal Anti-DNP antibody, Mouse IgG1 Avitag Isotype Control (Cat. No. DNP-BM190) and positive control antibody (Cat. No. FM3-BY54) respectively, washed and then followed by PE-SA and analyzed with FACS (QC tested).;5e5 of PBMCs were stained with 100 μL of 1:50 dilution (2 μL stock solution in 100 μL FACS
Catalog No. 50-210-7500
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.