Learn More
Lonza™ BioWhittaker™ Dulbecco's Phosphate Buffered Saline (1X) w/o Calcium and Magnesium
Description
- Serves as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time
- Manufactured in accordance with cGMP regulations and are cell culture tested to assure compatibility
Specifications
Specifications
| Content And Storage | 15° to 30°C |
| Product Type | Dulbecco's Phosphate Buffered Saline |
| Without Additives | No Calcium, No Magnesium |
| pH | 7 to 7.6 |
| Quantity | 500 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.