Learn More
Lonza™ BioWhittaker™ Dulbecco's Phosphate Buffered Saline (1X) w/o Calcium and Magnesium
Description
- Serves as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time
- Manufactured in accordance with cGMP regulations and are cell culture tested to assure compatibility
Specifications
Specifications
| Product Type | Dulbecco's Phosphate Buffered Saline |
| Without Additives | No Calcium, No Magnesium |
| pH | 7 to 7.6 |
| Quantity | 12 x 500 mL |