Learn More
Lonza™ BioWhittaker™ Phosphate Buffered Solution (1X)
Description
- Wash solution
- Manufactured in accordance with cGMP regulations
- Cell culture tested to assure compatibility
Specifications
Specifications
| Chemical Name or Material | Phosphate Buffered Saline (PBS) |
| pH | 7.3 to 7.5 |
| Quantity | 500 mL |
| Sterility | Sterile |
| Includes | No Calcium, No Magnesium |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.