Learn More
Sartorius Biowelder TC Disposable Blades in Box (1 box of 50 blades)
Supplier:  Sartorius 16389012
Encompass
The BioWelder TC is a fully automated welder for connecting thermoplastic tubing in a sterile welding operation. This new innovating technology allows sterile connection of dry or liquid filled TPE tubing up to 1 inch outer diameter. The BioWelder TC produces TPE tubing welding through a thermal connection and ensures an extraordinary level of stability and guarantees a sterile connection. The BioWelder TC is used to connect thermoplastic tubing such as C-Flex, AdvantaFlex, SaniPure BDF(TM) and PharMed BPT, used on disposable bags or bag assemblies within all biopharmaceutical manufacturing processes.
Catalog No. NC0853633