Learn More
Cayman Chemical Birinapant, 1260251-31-7, 5 mg
SDP
Supplier:  Cayman Chemical 196995
Encompass_Preferred
Molecular Formula C42H56F2N8O6, Purity ≥95%, Formula Weight 806.9, Synonyms: (2S,2'S)-N,N'-[(6,6'-difluoro[2,2'-bi-1H-indole]-3,3'-diyl)bis[methylene[(2R,4S)-4-hydroxy-2,1-pyrrolidinediyl][(1S)-1-ethyl-2-oxo-2,1-ethanediyl]]]bis[2-(methylamino)]-propanamide, TL-32711; A bivalent Smac mimetic that antagonizes cIAP1, cIAP2, and XIAP at nanomolar concentrations; causes caspase activation, PARP cleavage, and NF-B activation in breast cancer cells.
Catalog No. NC1344571
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.