Learn More
TARGETMOL CHEMICALS INC BIRM 271 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T776385MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. BIRM 271 is a novel arachidonic acid release inhibitor that blocks leukotriene B4 and platelet-activating factor biosynthesis in human neutrophils. BIRM 271 and BIRM 270 are enantiomers that inhibit the production of leukotriene B4 with IC50 of 40 nM. purity: 99%
Catalog No. 50-257-9127