Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2,2,2-trifluoroethyl) Carbonate 98.0+%, TCI America™
SDP

Catalog No. B470325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B470325G 25 g
B47035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B470325G Supplier TCI America Supplier No. B470325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1513-87-7
Molecular Formula C5H4F6O3
Molecular Weight (g/mol) 226.074
MDL Number MFCD11857845
InChI Key WLLOZRDOFANZMZ-UHFFFAOYSA-N
Synonym Carbonic Acid Bis(2,2,2-trifluoroethyl) Ester
PubChem CID 11447463
IUPAC Name bis(2,2,2-trifluoroethyl) carbonate
SMILES C(C(F)(F)F)OC(=O)OCC(F)(F)F

Specifications

Color Yellow
Boiling Point 118°C
Quantity 25 g
UN Number 3272
Formula Weight 226.07
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis(2,2,2-trifluoroethyl) Carbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.