Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis[2-(2-benzothiazolyl)phenolato]zinc(II) 98.0+%, TCI America™
SDP

Catalog No. B20771G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20771G 1 g
B20775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B20771G Supplier TCI America Supplier No. B20771G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 58280-31-2
Molecular Formula C26H18N2O2S2Zn
Molecular Weight (g/mol) 519.942
MDL Number MFCD03844766
InChI Key MPDFTJIWQKPYAF-YJCYQSLBSA-N
PubChem CID 50878549
IUPAC Name (6E)-6-(3H-1,3-benzothiazol-2-ylidene)cyclohexa-2,4-dien-1-one;zinc
SMILES C1=CC=C2C(=C1)NC(=C3C=CC=CC3=O)S2.C1=CC=C2C(=C1)NC(=C3C=CC=CC3=O)S2.[Zn]

Specifications

Quantity 1 g
Formula Weight 517.93
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis[2-(2-benzothiazolyl)phenolato]zinc(II)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.