Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2,2-diethoxyethyl) Disulfide 90.0+%, TCI America™
SDP

Catalog No. B28065G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28065G 5 g
B28061G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B28065G Supplier TCI America Supplier No. B28065G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 76505-71-0
Molecular Formula C12H26O4S2
Molecular Weight (g/mol) 298.46
MDL Number MFCD07357407
InChI Key XJIKEAQNTPCOLU-UHFFFAOYSA-N
PubChem CID 10990187
IUPAC Name 2-[(2,2-diethoxyethyl)disulfanyl]-1,1-diethoxyethane
SMILES CCOC(CSSCC(OCC)OCC)OCC

Specifications

Boiling Point 155°C
Quantity 5 g
Formula Weight 298.46
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis(2,2-diethoxyethyl) Disulfide
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.