Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2,4-pentanedionato)manganese(II) Dihydrate 97.0+%, TCI America™
SDP

Catalog No. M0042500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0042500G 500 g
M004225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0042500G Supplier TCI America Supplier No. M0042500G

Chemical Identifiers

CAS 14024-58-9
Molecular Formula C10H14MnO4
Molecular Weight (g/mol) 253.16
MDL Number MFCD00000022 MFCD09998212
InChI Key ZQZQURFYFJBOCE-FDGPNNRMSA-L
Synonym bis 4-hydroxypent-3-en-2-one dihydrate manganese
PubChem CID 54669727
IUPAC Name manganese(2+) bis((2Z)-4-oxopent-2-en-2-olate)
SMILES [Mn++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Color Yellow
Quantity 500 g
Formula Weight 253.16
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(2,4-pentanedionato)manganese(II) Dihydrate
EINECSNumber (2)-2142
RTECSNumber OO9350000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.