Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2-carboxyethyl) Isocyanurate 98.0+%, TCI America™
SDP

Catalog No. I058425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I058425G 25 g
I0584500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I058425G Supplier TCI America Supplier No. I058425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2904-40-7
Molecular Formula C9H11N3O7
Molecular Weight (g/mol) 273.201
MDL Number MFCD04038422
InChI Key JCUMQTAAEUDUPK-UHFFFAOYSA-N
Synonym Isocyanuric Acid Bis(2-carboxyethyl) Ester
PubChem CID 223763
IUPAC Name 3-[3-(2-carboxyethyl)-2,4,6-trioxo-1,3,5-triazinan-1-yl]propanoic acid
SMILES C(CN1C(=O)NC(=O)N(C1=O)CCC(=O)O)C(=O)O

Specifications

Melting Point 289°C
Color White
Quantity 25 g
Formula Weight 273.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(2-carboxyethyl) Isocyanurate
EINECSNumber (5)-1063
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.