Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis[2-carboxyethylgermanium(IV)] Sesquioxide 98.0+%, TCI America™
SDP

Catalog No. B13675G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B13675G 5 g
B136725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B13675G Supplier TCI America Supplier No. B13675G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 12758-40-6
Molecular Formula C6H10Ge2O7
Molecular Weight (g/mol) 339.399
MDL Number MFCD00049319
InChI Key XEABSBMNTNXEJM-UHFFFAOYSA-N
Synonym propagermanium, proxigermanium, carboxyethylgermanium sesquioxide, 14c-proxigermanium, 2-carboxyethylgermasesquioxane, dipropanoic acid germanium sequioxide, 2-carboxyethyl germanium sesquioxide, bis 2-carboxyethylgermanium sesquioxide, ccris 5469, carboxylethylgermanium sesquioxide
PubChem CID 83030
ChEBI CHEBI:32060
IUPAC Name 3-[[2-carboxyethyl(oxo)germyl]oxy-oxogermyl]propanoic acid
SMILES C(C[Ge](=O)O[Ge](=O)CCC(=O)O)C(=O)O

Specifications

Melting Point 320°C
Color White-Yellow
Quantity 5 g
Formula Weight 339.40
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis[2-carboxyethylgermanium(IV)] Sesquioxide
RTECSNumber UA2459750
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.