Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2-chloroethyl)amine hydrochloride, 98%

Catalog No. AAA131180B
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA131180B 1000 g
AAA1311818 50 g
AAA1311830 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA131180B Supplier Thermo Scientific Chemicals Supplier No. A131180B
Add to Cart

CAS: 821-48-7 | C4H10Cl3N | 178.481 g/mol

Bis(2-chloroethyl)amine hydrochloride is used as an intermediate in chemicals synthesis and building block for piperazine derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(2-chloroethyl)amine hydrochloride is used as an intermediate in chemicals synthesis and building block for piperazine derivatives.

Solubility
Partly miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 821-48-7
Molecular Formula C4H10Cl3N
Molecular Weight (g/mol) 178.481
MDL Number MFCD00012515
InChI Key YMDZDFSUDFLGMX-UHFFFAOYSA-N
Synonym bis 2-chloroethyl amine hydrochloride, 2,2'-dichlorodiethylamine hydrochloride, 2-chloro-n-2-chloroethyl ethanamine hydrochloride, nor-lost hydrochlorid, nor-hn2, ethanamine, 2-chloro-n-2-chloroethyl-, hydrochloride, n,n-bis 2-chloroethyl amine hydrochloride, di-2-chloroethyl amine hydrochloride, 2,2-dichlorodiethylamine hydrochloride, bis-2-chloroethyl amine hydrochloride
PubChem CID 522769
IUPAC Name 2-chloro-N-(2-chloroethyl)ethanamine;hydrochloride
SMILES C(CCl)NCCCl.Cl

Specifications

Melting Point 211°C to 215°C
Quantity 1000 g
UN Number UN2923
Beilstein 3550356
Solubility Information Partly miscible in water.
Formula Weight 178.49
Percent Purity 98%
Chemical Name or Material Bis(2-chloroethyl)amine hydrochloride
Hazard Category H302-H314-H335-H340
Hazard Statement GHS H Statement
H331-H340-H350-H314-H318-H302
Toxic if inhaled.
May cause genetic defects.
May cause cancer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
EINECSNumber 212-479-5
RTECSNumber IA1225000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.