Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2-ethylhexyl) adipate, 99%, MilliporeSigma™ Supelco™

Catalog No. 111012337 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2337 1 L
11-101-2338 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-2337 Supplier MilliporeSigma Supelco Supplier No. 5251971L
Add to Cart
Add to Cart

Chemical Identifiers

MDL Number MFCD00009496
Synonym Adipic acid di(2-ethylhexyl) ester; DOA

Specifications

Melting Point <-70°C
Density 0.925 g/mL (at 20°C)
CAS 103-23-1
Boiling Point 175°C (2 mmHg)
Flash Point 196°C
Packaging Glass Bottle
Linear Formula [-CH2 CH2 CO2 CH2 CH(C2H5)(CH2)3 CH3]2
Quantity 1 L
Formula Weight 370.57
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.