Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2-ethylhexyl) Isophthalate 98.0+%, TCI America™
SDP

Catalog No. B430225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B430225G 25 g
B4302500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B430225G Supplier TCI America Supplier No. B430225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 137-89-3
Molecular Formula C24H38O4
Molecular Weight (g/mol) 390.564
MDL Number MFCD00152284
InChI Key WXZOXVVKILCOPG-UHFFFAOYSA-N
Synonym Isophthalic Acid Bis(2-ethylhexyl) Ester
PubChem CID 8733
IUPAC Name bis(2-ethylhexyl) benzene-1,3-dicarboxylate
SMILES CCCCC(CC)COC(=O)C1=CC(=CC=C1)C(=O)OCC(CC)CCCC

Specifications

Color Colorless
Boiling Point 211°C
Quantity 25 g
Formula Weight 390.56
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis(2-ethylhexyl) Isophthalate
EINECSNumber (3)-1319
RTECSNumber NT2050000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.