Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2-iodoethyl) Ether (stabilized with Copper chip) 98.0+%, TCI America™
SDP

Catalog No. B54395G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B54395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B54395G Supplier TCI America Supplier No. B54395G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 34270-90-1
Molecular Formula C4H8I2O
Molecular Weight (g/mol) 325.92
MDL Number MFCD00092250
InChI Key XWYDQETVMJZUOJ-UHFFFAOYSA-N
Synonym 2,2′-Diiododiethyl Ether, 2-Iodoethyl Ether
PubChem CID 3430779
IUPAC Name 1-iodo-2-(2-iodoethoxy)ethane
SMILES ICCOCCI

Specifications

Boiling Point 124°C
Quantity 5 g
Formula Weight 325.92
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis(2-iodoethyl) Ether (stabilized with Copper chip)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.