Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis[(2R,5R)-1-(2-[(2R,5R)-2,5-dimethyl-1-phospholanyl]phenyl)-2,5-dimethylphospholane 1-oxide]copper(I) triflate, 97%

Catalog No. AAH63492ME
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63492ME 500 mg
AAH63492MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH63492ME Supplier Thermo Scientific Chemicals Supplier No. H63492ME

missing translation for 'validMainframeMsg'

CAS: 874013-62-4 | C37H56CuF3O5P4S | 857.346 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 874013-62-4
Molecular Formula C37H56CuF3O5P4S
Molecular Weight (g/mol) 857.346
MDL Number MFCD17018797
InChI Key JGVWSKIVCRMXGT-YCIFJGFLSA-M
Synonym bis 1-2r,5r-2,5-dimethylphospholanyl-2-2r,5r-2,5-dimethylphospholanyl-oxide benzene cu i trifluoromethanesulfonate
PubChem CID 11707733
IUPAC Name copper(1+);(2R,5R)-1-[2-[(2R,5R)-2,5-dimethylphospholan-1-yl]phenyl]-2,5-dimethyl-1$l^{5}-phospholane 1-oxide;trifluoromethanesulfonate
SMILES CC1CCC(P1C2=CC=CC=C2P3(=O)C(CCC3C)C)C.CC1CCC(P1C2=CC=CC=C2P3(=O)C(CCC3C)C)C.C(F)(F)(F)S(=O)(=O)[O-].[Cu+]

Specifications

Color Brown to White
Quantity 500 mg
Sensitivity Air Sensitive
Formula Weight 857.34
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Bis[(2R,5R)-1-(2-[(2R,5R)-2,5-dimethyl-1-phospholanyl]phenyl)-2,5-dimethylphospholane 1-oxide]copper
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.