Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam Bis(3,5-dibromosalicyl) Fumarate, Hemoglobin Acylating Agent, 1 g
Description
- Molecular weight: 671.9 Da
- Purity: >93%
- Description: Potent hemoglobin acylating agent that cross-links beta chains of hemoglobin
- Function: Acylating and anti-sickling agent
- Activity: Active in vitro and in vivo
- Chemical name: Bis(3,5-dibromosalicyl) fumarate
Specifications
Specifications
| Target Molecule | Hemoglobin |
| Substrate Properties | Chemical Substrate |
| Shipping Condition | Ambient |
| Form | Solid |
| Buffer | 100% Bis(3,5-dibromosalicyl) fumarate |
| Content And Storage | Store at -20°C. |
| Formulation | 100% Bis(3,5-dibromosalicyl) fumarate |
| Quantity | 1 g |
| For Use With (Application) | Hemoglobin acylation, anti-sickling research |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?