Learn More
Abcam Bis(3,5-dibromosalicyl) Fumarate, Hemoglobin Acylating Agent, 100 mg
Description
- Molecular Weight: 671.9 Da
- Purity: >93%
- Description: Potent hemoglobin acylating agent that cross-links beta chains of hemoglobin; acts as an acylating and anti-sickling agent
- Activity: Active in vitro and in vivo
- Composition: 100% Bis(3,5-dibromosalicyl) fumarate
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Chemical Substrate |
| Target Molecule | Hemoglobin |
| Buffer | 100% Bis(3,5-dibromosalicyl) fumarate |
| Content And Storage | Store at -20°C. |
| Shipping Condition | Ambient |
| Substrate | Hemoglobin |
| Formulation | 100% Bis(3,5-dibromosalicyl) fumarate |
| Quantity | 100 mg |
| For Use With (Application) | Hemoglobin acylation, anti-sickling research |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.