Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Bis(3,5-dibromosalicyl) Fumarate, Hemoglobin Acylating Agent, 671.9 Da, 5 g

Catalog No. 502706160
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6160 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6160 Supplier Abcam Supplier No. AB1428365G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Potent hemoglobin acylating and anti-sickling agent that cross-links beta chains of hemoglobin.

  • Molecular weight: 671.9 Da
  • Purity: >93%
  • Potent hemoglobin acylating agent
  • Cross-links beta chains of hemoglobin
  • Acylating and anti-sickling agent
  • Active in vitro and in vivo
  • CAS number: 71337-53-6

Specifications

Substrate Properties Chemical Substrate
Target Molecule Hemoglobin
Buffer 100% Bis(3,5-dibromosalicyl) fumarate
Content And Storage Store at -20°C.
Shipping Condition Ambient
Substrate Hemoglobin
Formulation 100% Bis(3,5-dibromosalicyl) fumarate
Quantity 5 g
For Use With (Application) Hemoglobin acylation, anti-sickling research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.