Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Bis(3,5-dibromosalicyl) Fumarate, Hemoglobin Acylating Agent, >93% Purity, 500 mg

Catalog No. 502706161
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6161 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6161 Supplier Abcam Supplier No. AB142836500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent hemoglobin acylating and anti-sickling agent that cross-links hemoglobin beta chains.

  • Molecular weight: 671.9 Da
  • Purity: >93%
  • Functional properties: Potent hemoglobin acylating agent; cross-links beta chains of hemoglobin; acts as an acylating and anti-sickling agent
  • Activity: Active in vitro and in vivo
  • Composition: 100% Bis(3,5-dibromosalicyl) fumarate

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Hemoglobin
Substrate Properties Chemical Substrate
Shipping Condition Ambient
Form Powder
Buffer 100% Bis(3,5-dibromosalicyl) fumarate
Content And Storage Store at -20°C.
Formulation 100% Bis(3,5-dibromosalicyl) fumarate
Quantity 500 mg
For Use With (Application) Hemoglobin acylating, anti-sickling
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.