Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(3-aminopropyl) Ether 98.0+%, TCI America™
SDP

Catalog No. B12685ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B12685ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B12685ML Supplier TCI America Supplier No. B12685ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2157-24-6
Molecular Formula C6H16N2O
Molecular Weight (g/mol) 132.207
MDL Number MFCD00059961
InChI Key KRPRVQWGKLEFKN-UHFFFAOYSA-N
Synonym 3,3′C-Oxydi(propylamine)
PubChem CID 12841412
IUPAC Name 3-(3-aminopropoxy)propan-1-amine
SMILES C(CN)COCCCN

Specifications

Color Colorless
Boiling Point 72°C
Quantity 5 mL
UN Number 3267
Formula Weight 132.21
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Bis(3-aminopropyl) Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.