Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(3-bromophenylboronic Acid) scyllo-Inositol Complex Dipotassium Pentahydrate, TCI America™
SDP

Catalog No. B45231G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45231G 1 g
B4523200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B45231G Supplier TCI America Supplier No. B45231G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1537876-31-5
Molecular Formula C18H24B2Br2K2O11
Molecular Weight (g/mol) 676.004
InChI Key PJGZRWPVEOHJHW-UHFFFAOYSA-N
PubChem CID 131674833
SMILES [B-]12(OC3C4C(O1)C5C(C3O[B-](O4)(O5)C6=CC(=CC=C6)Br)O2)C7=CC(=CC=C7)Br.O.O.O.O.O.[K+].[K+]

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 585.93
Physical Form Crystalline Powder
Chemical Name or Material Bis(3-bromophenylboronic Acid) scyllo-Inositol Complex Dipotassium Pentahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.