Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(3-ethyl-5-methyl-4-maleimidophenyl)methane 98.0+%, TCI America™
SDP

Catalog No. B1971100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1971100G 100 g
B197125G 25 g
B1971500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B1971100G Supplier TCI America Supplier No. B1971100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 105391-33-1
Molecular Formula C27H26N2O4
Molecular Weight (g/mol) 442.515
MDL Number MFCD00467754
InChI Key YNSSPVZNXLACMW-UHFFFAOYSA-N
Synonym Methylenebis(3-ethyl-5-methyl-4-maleimidobenzene)
PubChem CID 10950302
IUPAC Name 1-[4-[[4-(2,5-dioxopyrrol-1-yl)-3-ethyl-5-methylphenyl]methyl]-2-ethyl-6-methylphenyl]pyrrole-2,5-dione
SMILES CCC1=CC(=CC(=C1N2C(=O)C=CC2=O)C)CC3=CC(=C(C(=C3)CC)N4C(=O)C=CC4=O)C

Specifications

Melting Point 165°C
Color White-Yellow
Quantity 100 g
Formula Weight 442.52
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Bis(3-ethyl-5-methyl-4-maleimidophenyl)methane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.