Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis[3-(trimethoxysilyl)propyl]amine 98.0+%, TCI America™
SDP

Catalog No. B2548100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2548100G 100 g
B254825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B2548100G Supplier TCI America Supplier No. B2548100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 82985-35-1
Molecular Formula C12H31NO6Si2
Molecular Weight (g/mol) 341.551
MDL Number MFCD00191835
InChI Key TZZGHGKTHXIOMN-UHFFFAOYSA-N
PubChem CID 157882
IUPAC Name 3-trimethoxysilyl-N-(3-trimethoxysilylpropyl)propan-1-amine
SMILES CO[Si](CCCNCCC[Si](OC)(OC)OC)(OC)OC

Specifications

Color Yellow
Boiling Point 152°C
Quantity 100 g
UN Number 3082
Formula Weight 341.55
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Bis[3-(trimethoxysilyl)propyl]amine
RTECSNumber TX2101000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.