Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(4-cyanophenyl) Ether 98.0+%, TCI America™
SDP

Catalog No. B279025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B279025G 25 g
B27905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B279025G Supplier TCI America Supplier No. B279025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6508-04-9
Molecular Formula C14H8N2O
Molecular Weight (g/mol) 220.23
InChI Key RSAUOQFEFINEDM-UHFFFAOYSA-N
PubChem CID 81004
IUPAC Name 4-(4-cyanophenoxy)benzonitrile
SMILES C1=CC(=CC=C1C#N)OC2=CC=C(C=C2)C#N

Specifications

Melting Point 184°C
Color White
Boiling Point 220°C
Quantity 25 g
UN Number 3439
Synonym 4,4′-Dicyanodiphenyl Ether, 4,4′-Oxydibenzonitrile
Formula Weight 220.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(4-cyanophenyl) Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.