Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(4-methoxyphenyl) Selenoxide 93.0+%, TCI America™
SDP

Catalog No. B13751G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B13751G 1 g
B13755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B13751G Supplier TCI America Supplier No. B13751G

Chemical Identifiers

Molecular Formula C14H14O3Se
Molecular Weight (g/mol) 309.234
MDL Number MFCD00059043
InChI Key SXEWETDJNQVWQY-UHFFFAOYSA-N
PubChem CID 11141388
IUPAC Name 1-methoxy-4-(4-methoxyphenyl)seleninylbenzene
SMILES COC1=CC=C(C=C1)[Se](=O)C2=CC=C(C=C2)OC

Specifications

Color White-Yellow
Quantity 1 g
UN Number 3283
Formula Weight 309.22
Percent Purity ≥93.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(4-methoxyphenyl) Selenoxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.