Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(4-nitrophenyl) Phosphate 98.0+%, TCI America™
SDP

Catalog No. B109810G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B109810G 10 g
B10981G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B109810G Supplier TCI America Supplier No. B109810G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 645-15-8
Molecular Formula C12H9N2O8P
Molecular Weight (g/mol) 340.18
MDL Number MFCD00007318
InChI Key MHSVUSZEHNVFKW-UHFFFAOYSA-N
Synonym Phosphoric Acid Bis(4-nitrophenyl) Ester
PubChem CID 255
ChEBI CHEBI:3122
IUPAC Name bis(4-nitrophenoxy)phosphinic acid
SMILES OP(=O)(OC1=CC=C(C=C1)[N+]([O-])=O)OC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 176°C
Color White
Quantity 10 g
Formula Weight 340.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(4-nitrophenyl) Phosphate [for Phosphodiesterase Substrate]
RTECSNumber TB8260000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.