Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

bis-Benzimide H-33342 Trihydrochloride, MP Biomedicals

Catalog No. ICN19030525 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19030525 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19030525 Supplier MP Biomedicals, Inc Supplier No. 0219030525

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 23491-52-3
Molecular Formula C27H28N6O
Molecular Weight (g/mol) 452.562
InChI Key PRDFBSVERLRRMY-UHFFFAOYSA-N
Synonym hoechst 33342, bisbenzimide, hoechst33342, unii-99kzs6cnzx, 99kzs6cnzx, 2'-4-ethoxyphenyl-5-4-methylpiperazin-1-yl-1h,1'h-2,5'-bibenzo d imidazole, 2'-4-ethoxyphenyl-5-4-methyl-1-piperazinyl-2,5'-bi-benzimidazole, 2'-4-ethoxyphenyl-5-4-methyl-1-piperazinyl-2,5'-bi-1h-benzimidazole, 2'-4'-ethoxyphenyl-5-4-methylpiperazin-1-yl-2,5'-bis-1h-benzimidazole trihydrochloride trihydrate, 2'-p-ethoxyphenyl-5-4-methyl-1-piperazinyl-2,5'-bibenzimidazole
PubChem CID 1464
ChEBI CHEBI:51232
IUPAC Name 2-(4-ethoxyphenyl)-6-[6-(4-methylpiperazin-1-yl)-1H-benzimidazol-2-yl]-1H-benzimidazole
SMILES CCOC1=CC=C(C=C1)C2=NC3=C(N2)C=C(C=C3)C4=NC5=C(N4)C=C(C=C5)N6CCN(CC6)C

Specifications

Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.