accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 23491-52-3

IUPAC Name: 2-(4-ethoxyphenyl)-6-[6-(4-methylpiperazin-1-yl)-1H-1,3-benzodiazol-2-yl]-1H-1,3-benzodiazole
Synonyms: bisbenzimide
Molecular Weight (g/mol): 452.56
Molecular Formula: C27H28N6O
InChi Key: PRDFBSVERLRRMY-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(C=C1)C1=NC2=CC=C(C=C2N1)C1=NC2=CC=C(C=C2N1)N1CCN(C)CC1
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)