Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

bis-Benzimide H-33342 trihydrochloride trihydrate, 98%

Catalog No. AAJ62134MC
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62134MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ62134MC Supplier Thermo Scientific Chemicals Supplier No. J62134MC
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 23491-52-3 | C27H28N6O | 452.562 g/mol

Stain for identifying specific chromosome banding patterns

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23491-52-3
Molecular Formula C27H28N6O
Molecular Weight (g/mol) 452.562
MDL Number MFCD00012679
InChI Key PRDFBSVERLRRMY-UHFFFAOYSA-N
Synonym hoechst 33342, bisbenzimide, hoechst33342, unii-99kzs6cnzx, 99kzs6cnzx, 2'-4-ethoxyphenyl-5-4-methylpiperazin-1-yl-1h,1'h-2,5'-bibenzo d imidazole, 2'-4-ethoxyphenyl-5-4-methyl-1-piperazinyl-2,5'-bi-benzimidazole, 2'-4-ethoxyphenyl-5-4-methyl-1-piperazinyl-2,5'-bi-1h-benzimidazole, 2'-4'-ethoxyphenyl-5-4-methylpiperazin-1-yl-2,5'-bis-1h-benzimidazole trihydrochloride trihydrate, 2'-p-ethoxyphenyl-5-4-methyl-1-piperazinyl-2,5'-bibenzimidazole
PubChem CID 1464
ChEBI CHEBI:51232
IUPAC Name 2-(4-ethoxyphenyl)-6-[6-(4-methylpiperazin-1-yl)-1H-benzimidazol-2-yl]-1H-benzimidazole
SMILES CCOC1=CC=C(C=C1)C2=NC3=C(N2)C=C(C=C3)C4=NC5=C(N4)C=C(C=C5)N6CCN(CC6)C

Specifications

Color Yellow to Green
Quantity 100 mg
Beilstein 4088183
Formula Weight 615.99 (561.94 Anhydrous)
Percent Purity 98%
Physical Form Powder
Chemical Name or Material bis-Benzimide H-33342 trihydrochloride trihydrate
Hazard Category Warning
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 245-691-1
RTECSNumber SM1140500
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.