Learn More
Strem, An Ascensus Company CAS# 1271-19-8. Bis(cyclopentadienyl)titanium dichloride, 99+% (Titanocene dichloride) MFCD00003723. 100G
SDP
Supplier:  Strem, An Ascensus Company 220200100G
Encompass_Preferred
CAS# 1271-19-8. 100G. Bis(cyclopentadienyl)titanium dichloride, 99+% (Titanocene dichloride). MFCD00003723. Molecular Formula: C10H10Cl2Ti Molecular Weight: 249.00. Color/Form: red xtl. Stability: air sensitive, moisture sensitive. Strem#: 22-0200 www.strem.com/catalog/v3/22-0200/
Catalog No. 50-901-10729
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.