Learn More
Bis(dibenzylideneacetone)palladium(0)
Description
Hydrogenation, isomerization, carbonylation, oxidation, C-C bond formation.Bis(dibenzylideneacetone)palladium(0) is used as a reagent for the preparation of allylic substituted cyclopentadienes. It acts as a homogeneous catalyst as well as in the alkylation of allyl acetates by various nucleophiles. Further, it is used in Suzuki reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 135°C (decomposition) |
| Color | Red to Brown |
| Quantity | 1 g |
| Sensitivity | Air and moisture sensitive |
| Solubility Information | Slightly soluble in CH2Cl2,CHCl3 and benzene.Soluble in dichloromethane,chloroform,1,2-dichloroethane,acetone,acetonitrile and benzene. Insoluble in water. |
| Formula Weight | 575 |
| Physical Form | Powder |
| Chemical Name or Material | Bis(dibenzylideneacetone) palladium(0) |
Safety and Handling
| Hazard Category | H317 |
| Precautionary Statement | P261-P272-P280g-P302+P352-P333+P313-P363-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.