Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(dimethylphosphino)methane, 98%

Catalog No. AA3927504
Change view
Click to view available options
Quantity:
0.5 g
2 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3927504 2 g
AA3927502 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3927504 Supplier Thermo Scientific Chemicals Supplier No. 03927504

CAS: 64065-08-3 | C5H14P2 | 136.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64065-08-3
Molecular Formula C5H14P2
Molecular Weight (g/mol) 136.12
MDL Number MFCD00058731
InChI Key MRNJHNUEBDGNEL-UHFFFAOYSA-N
Synonym bis dimethylphosphino methane, phosphine,1,1'-methylenebis 1,1-dimethyl, dmpm, acmc-1b3lc, dimethylphosphanylmethyl dimethyl phosphane, dimethylphosphanyl methyl dimethylphosphane, dimethylphosphino methyl dimethyl phosphine, dimethylphosphino methyl dimethyl phosphine #
PubChem CID 522114
SMILES CP(C)CP(C)C

Specifications

Color Colorless
Density 0.86 g/mL
Boiling Point 42°C (12 mmHg)
Linear Formula (CH3)2PCH2P(CH3)2
Quantity 2 g
UN Number UN2845
Sensitivity Air and moisture sensitive
IUPAC Name dimethylphosphanylmethyl(dimethyl)phosphane
Formula Weight 136.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Bis(dimethylphosphino)methane
Show More Show Less
Hazard Category H250-H252-H315-H319-H335
Hazard Statement GHS H Statement
H250-H252-H315-H319-H335
Catches fire spontaneously if exposed to air.
Self-heating in large quantities
may catch fire.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P222-P235+P410-P261-P264b-P271-P280-P302+P334-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC LIQUIDS, ORGANIC, N.O.S.
TSCA No
storageNote1
  • Packaged under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.