Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bis-Mal-Dpeg™11

Catalog No. QBD10397 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 mg
1000 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
QBD10397 1000 mg
QBD10397MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. QBD10397 Supplier Sigma Aldrich Supplier No. QBD103971000MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Bis-MAL-dPEG ™11 is useful for producing dimeric molecules after reaction with free thiol groups.dPEG is a registered trademark of Quanta BioDesign

Specifications

CAS 1334178-05-0
Molecular Formula C38H62N4O17
Linear Formula C38H62N4O17
MDL Number MFCD13184981
Quantity 1000 mg
Molecular Weight (g/mol) 846.92 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.