Learn More
Enzo Life Sciences bis(Methylthio)gliotoxin (1mg). CAS: 74149-38-5
SDP
Supplier:  Enzo Life Sciences BMLL1220001
Encompass_Preferred
Inhibits PAF-induced platelet aggregation (IC50=8.4 µM) and collagen-induced aggregation (IC50=84.2 µM) with no effect on arachidonic acid or ADP induced aggregation. Inhibits PAF-induced bronchoconstriction in vivo in the guinea-pig (69% at 1.0 mg/kg). Alternative name: FR-49175. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (25mg/ml) and ethanol (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-1134
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.