Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(N,N-dimethylacetamide) Hydrogen Dibromobromate 97.0+%, TCI America™
SDP

Catalog No. B2509250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2509250G 250 g
B250925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B2509250G Supplier TCI America Supplier No. B2509250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 75381-80-5
Molecular Formula C8H19Br3N2O2
Synonym Bis(N,N-dimethylacetamide) Hydrogen Tribromide

Specifications

Melting Point 92°C
Quantity 250 g
Formula Weight 414.96
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(N,N-dimethylacetamide) Hydrogen Dibromobromate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.