Learn More
Bis(n-propylcyclopentadienyl)magnesium, 98+%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Density | 0.94g/mL |
| Packaging | Ampule |
| Quantity | 1 g |
| UN Number | UN3391 |
| Sensitivity | Air & Moisture Sensitive |
| Formula Weight | 238.66 |
| Percent Purity | ≥98% |
| Physical Form | Crystalline |
| Chemical Name or Material | Bis(n-propylcyclopentadienyl)magnesium |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement H250-H252 Catches fire spontaneously if exposed to air. Self-heating in large quantities may catch fire. |
| Precautionary Statement | GHS P Statement P210-P222-P280-P370+P378a-P405-P422a Keep away from heat/sparks/open flames/hot surfaces. No smoking. Do not allow contact with air. Wear protective gloves/protective clothing/eye protection/face protection. In case of fire: Use for extinction: CO2, powder or water spray. Store locked up. Store contents under inert gas. |
| DOTInformation | Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, SOLID, PYROPHORIC |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.