Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(neopentyl Glycolato)diboron 98.0+%, TCI America™
SDP

Catalog No. B225425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B225425G 25 g
B22541G 1 g
B22545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B225425G Supplier TCI America Supplier No. B225425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 201733-56-4
Molecular Formula C10H20B2O4
Molecular Weight (g/mol) 225.89
MDL Number MFCD02093062
InChI Key MDNDJMCSXOXBFZ-UHFFFAOYSA-N
Synonym bis neopentyl glycolato diboron, bis neopentylglycolato diboron, 5,5,5',5'-tetramethyl-2,2'-bi 1,3,2-dioxaborinane, 2-5,5-dimethyl-1,3,2-dioxaborinan-2-yl-5,5-dimethyl-1,3,2-dioxaborinane, 5,5,5',5'-tetramethyl-2,2'-bi-1,3,2-dioxaborinane, 2,2'-bi-1,3,2-dioxaborinane, 5,5,5',5'-tetramethyl, bis neopentyl glycolato diborane, bis 2,2-dimethyl-1,3-propanediolato diboron, 5,5,5',5'-tetramethyl-2,2' bi 1,3,2 dioxaborinanyl
PubChem CID 2734316
IUPAC Name 2-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)-5,5-dimethyl-1,3,2-dioxaborinane
SMILES CC1(C)COB(OC1)B1OCC(C)(C)CO1

Specifications

Melting Point 192°C
Color White-Yellow
Quantity 25 g
Formula Weight 225.89
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(neopentyl Glycolato)diboron
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.