Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(pyridine)iodonium tetrafluoroborate, 97%

Catalog No. AAL1938503
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1938503 1 g
AAL19385MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1938503 Supplier Thermo Scientific Chemicals Supplier No. L1938503
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 15656-28-7 | C10H10BF4IN2 | 371.91 g/mol

Bis(pyridine)iodonium tetrafluoroborate is used as mild iodinating and oxidizing reagent. It is also useful for selective iodination of free phenolic groups in tyrosine residues during solid-phase synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(pyridine)iodonium tetrafluoroborate is used as mild iodinating and oxidizing reagent. It is also useful for selective iodination of free phenolic groups in tyrosine residues during solid-phase synthesis.

Solubility
Hydrolyzes in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15656-28-7
Molecular Formula C10H10BF4IN2
Molecular Weight (g/mol) 371.91
MDL Number MFCD03703393
InChI Key BMDSRCBKJZCUBH-UHFFFAOYSA-N
Synonym bis pyridine iodonium tetrafluoroborate, barluenga's reagent, bis pyridine iodonium i tetrafluoroborate, iodanium pyridine tetrafluoroborate, iodonium pyridine tetrafluoroborate, bispyridine iodonium tetrafluoroborate, bis pyridin iodonium tetrafluoroborate, iodium bis pyridine tetrafluoroborate, bis pyridine-iodonium tetrafluoroborate
PubChem CID 10883201
SMILES F[B-](F)(F)F.[I-]([N+]1=CC=CC=C1)[N+]1=CC=CC=C1

Specifications

Melting Point ∽160°C (decomposition)
Odor Odorless
Quantity 1 g
Beilstein 4284739
Sensitivity Light and moisture sensitive
Solubility Information Hydrolyzes in water.
IUPAC Name bis(pyridin-1-ium-1-yl)iodanuide; tetrafluoroboranuide
Formula Weight 371.91
Percent Purity 97%
Chemical Name or Material Bis(pyridine)iodonium tetrafluoroborate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.