Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(tert-butylcyclopentadienyl)titanium dichloride, Thermo Scientific™

Catalog No. AC347860010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC347860010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC347860010 Supplier Thermo Scientific Chemicals Supplier No. 347860010

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 79269-71-9
Molecular Formula C18H26Cl2Ti
Molecular Weight (g/mol) 361.17

Specifications

Melting Point 233°C
Packaging Glass bottle
Quantity 1 g
Formula Weight 361.17
Chemical Name or Material Bis(tert-butylcyclopentadienyl)titanium dichloride
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.