Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(tetrabutylammonium) Dihydrogen Pyrophosphate 96.0+%, TCI America™
SDP

Catalog No. B54401G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B54401G 1 g
B5440200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B54401G Supplier TCI America Supplier No. B54401G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 857447-79-1
Molecular Formula C32H74N2O7P2
Molecular Weight (g/mol) 660.899
InChI Key WPGBCELIUCBRKN-UHFFFAOYSA-L
Synonym Bis(tetrabutylammonium) Dihydrogen Diphosphate
PubChem CID 11342793
IUPAC Name [hydroxy(oxido)phosphoryl] hydrogen phosphate;tetrabutylazanium
SMILES CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC.OP(=O)([O-])OP(=O)(O)[O-]

Specifications

Color White
Quantity 1 g
Formula Weight 660.90
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(tetrabutylammonium) Dihydrogen Pyrophosphate
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.