Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(trifluoromethanesulfonyl)imide 99.0+%, TCI America™
SDP

Catalog No. B254125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B254125G 25 g
B25415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B254125G Supplier TCI America Supplier No. B254125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 82113-65-3
Molecular Formula C2HF6NO4S2
Molecular Weight (g/mol) 281.143
MDL Number MFCD00214154
InChI Key ZXMGHDIOOHOAAE-UHFFFAOYSA-N
Synonym 1,1,1-trifluoro-n-trifluoromethyl sulfonyl methanesulfonamide, trifluoromethanesulfonimide, unii-c7r849vm9l, bis trifluoromethanesulfonyl imide, bistrifluoromethanesulfonimide, 1,1,1-trifluoro-n-trifluoromethanesulfonylmethanesulfonamide, 1,1,1-trifluoro-n-trifluoromethylsulfonyl methanesulfonamide, bis trifluoromethane sulfonimide, n,n-bis trifluoromethanesulfonyl imide, n,n-bis trifluoromethylsulphonyl amine
PubChem CID 157857
IUPAC Name 1,1,1-trifluoro-N-(trifluoromethylsulfonyl)methanesulfonamide
SMILES C(F)(F)(F)S(=O)(=O)NS(=O)(=O)C(F)(F)F

Specifications

Melting Point 56°C
Color White
Boiling Point 92°C
Quantity 25 g
UN Number 3261
Formula Weight 281.14
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(trifluoromethanesulfonyl)imide
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.