Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(triphenylphosphine)palladium(II) chloride, 98%

Catalog No. AC299250250
Change view
Click to view available options
Quantity:
2.5 g
5 g
25 g
100 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC299250250 25 g Glass bottle
AC299250025 2.5 g Glass bottle
AC299250050 5 g Glass bottle
AC299251000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC299250250 Supplier Thermo Scientific Chemicals Supplier No. 299250250
Add to Cart

CAS: 13965-03-2 | C36H30Cl2P2Pd | 701.90 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13965-03-2
Molecular Formula C36H30Cl2P2Pd
Molecular Weight (g/mol) 701.90
MDL Number MFCD00009593
InChI Key YNHIGQDRGKUECZ-UHFFFAOYSA-L
Synonym bis triphenylphosphine palladium ii dichloride, bis triphenylphosphine palladium ii chloride, dichlorobis triphenylphosphine palladium, bis triphenylphosphine palladium chloride, bis triphenylphosphine palladiuml ii dichloride, palladium 2+ ; triphenylphosphane; dichloride, trans-dichlorobis triphenylphosphine palladium ii 1g
PubChem CID 131664180
IUPAC Name ethane;methane;palladium(2+);triphenylphosphane;dichloride
SMILES [Cl-].[Cl-].[Pd++].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 260°C
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 14.8 to 15.5% (Pd)
Packaging Glass bottle
Linear Formula [(C6H5)3P]2PdCl2
Quantity 25 g
Fieser 06,59; 07,95; 08,151; 17,111
Solubility Information Solubility in water: insoluble
Formula Weight 701.89
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Bis(triphenylphosphine)palladium(II) chloride
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
May cause long lasting harmful effects to aquatic life.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
EINECSNumber 237-744-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.