Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(triphenylphosphine)palladium(II) Dichloride 98.0+%, TCI America™
SDP

Catalog No. B166725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B166725G 25 g
B16671G 1 g
B16675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B166725G Supplier TCI America Supplier No. B166725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13965-03-2
Molecular Formula C36H30Cl2P2Pd
Molecular Weight (g/mol) 701.90
MDL Number MFCD00009593
InChI Key YNHIGQDRGKUECZ-UHFFFAOYSA-L
Synonym bis triphenylphosphine palladium ii dichloride, bis triphenylphosphine palladium ii chloride, dichlorobis triphenylphosphine palladium, bis triphenylphosphine palladium chloride, bis triphenylphosphine palladiuml ii dichloride, palladium 2+ ; triphenylphosphane; dichloride, trans-dichlorobis triphenylphosphine palladium ii 1g
PubChem CID 131664180
IUPAC Name palladium(2+) bis(triphenylphosphane) dichloride
SMILES [Cl-].[Cl-].[Pd++].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 25 g
Formula Weight 701.90
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(triphenylphosphine)palladium(II) Dichloride
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.